Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

α-Benzoinoxime ≥98%

Supplier: ACROS ORGANICS MS

Synonyms: α-Benzoin oxime

AC10553-1000 AC10553-0250 AC10553-5000
200018-074EA 121.85 USD InStock
200018-074 200036-494 103508-412
α-Benzoinoxime ≥98%
α-Benzoinoxime


Formula: C₁₄H₁₃NO₂
MW: 227.26 g/mol
Melting Pt: 150…155 °C
Storage Temperature: Ambient
MDL Number: MFCD00004501
CAS Number: 441-38-3
EINECS: 207-127-2
Order Now
Specification Test Results
Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 150°C to 155°C
HPLC ≥97.5 %


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty