3-Amino-1-propanesulfonic acid 95%

Warning

Synonyms: Homotaurine3-Aminopropane-1-sulfonic acidTramiprosate3APS3-Aminopropane-1-sulphonic acid

AC10443-0050 AC10443-0010
103465-208EA 109 USD
103465-208 200016-924
3-Amino-1-propanesulfonic acid 95%
3-Amino-1-propanesulfonic acid
Formula: C₃H₉NO₃S
MW: 139.17 g/mol
Melting Pt: >270 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00008225
CAS Number: 3687-18-1

Order Now

Specification Test Results

Appearance White to light blue Crystalline powder
Infrared spectrum Conforms
Melting point ≥270 °C
Titration with NaOH ≥94.0 %
Phosphate (PO4) ≤3.5 % (by acid extraction)
Calcium (Ca) ≤200 ppm (by acid extraction)
Iron (Fe) ≤150 ppm (by acid extraction)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR