1-Aminopiperidine 97%

Danger

AC10441-0250 AC10441-1000 AC10441-0100
200053-712EA 830 USD
200053-712 200053-714 200053-716
1-Aminopiperidine 97%
1-Aminopiperidine
Formula: C₅H₁₂N₂
Density: 0.928 g/cm³
Flash Pt: 36 °C
Storage Temperature: Ambient
MDL Number: MFCD00006489
CAS Number: 2213-43-6
EINECS: 218-666-8
UN: 2920
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4735 to 1.4775 (20 °C, 589 nm)
GC ≥96.0 %
Calcium (Ca) ≤200 ppm
Copper (Cu) ≤10 ppm
Iron (Fe) ≤200 ppm
Insoluble matter ≤1 % (in acid)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR