2-Aminoethyl hydrogen sulfate 99%

AC10364-1000 AC10364-0025 AC10364-5000
200028-520EA 319 USD
200028-520 103508-648 200034-620
2-Aminoethyl hydrogen sulfate 99%
2-Aminoethyl hydrogen sulphate
Formula: C₂H₇NO₄S
MW: 141.1500 g/mol
Melting Pt: 277…280 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00008179
CAS Number: 926-39-6
EINECS: 213-135-7

Order Now

Specification Test Results

Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Titration with NaOH ≥98.5 %
Clarity of solution Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR