ɑ-Amino-γ-butyrolactone hydrobromide 99%

Warning

Synonyms: DL-Homoserine lactone hydrobromide

AC10329-0100 AC10329-0500 AC10329-0025
103508-628EA 362 USD
103508-628 103508-494 200011-080
ɑ-Amino-γ-butyrolactone hydrobromide 99%
alpha-Amino-gamma-butyrolactone hydrobromide
Formula: C₄H₇NO₂
MW: 182.02 g/mol
Melting Pt: 221 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012723
CAS Number: 6305-38-0

Order Now

Specification Test Results

Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Titration Argentometric ≥98.5 %
Dye content ≥65 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR