Aminoacetaldehyde diethyl acetal 99%

Danger

Synonyms: 2,2-Diethoxyethanamine2-Aminoacetaldehyde diethylacetal

AC10307-2500 AC10307-0500 AC10307-0050
103529-966EA 364 USD
103529-966 200036-538 200042-560
Aminoacetaldehyde diethyl acetal 99%
Aminoacetaldehyde diethyl acetal
Formula: C₆H₁₅NO₂
MW: 133.19 g/mol
Boiling Pt: 162…163 °C (760 mmHg)
Density: 0.916 g/cm³
Flash Pt: 45 °C
Storage Temperature: Ambient
MDL Number: MFCD00008136
CAS Number: 645-36-3
EINECS: 211-439-4
UN: 2733
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4160 to 1.4180 (20 °C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR