(±)-Allyl-2,3-epoxypropyl ether 99+%

Danger

Synonyms: Allyl glycidyl ether

AC10294-1000 AC10294-0025 AC10294-5000
200005-678EA 78.3 USD
200005-678 200018-038 103524-034
(±)-Allyl-2,3-epoxypropyl ether 99+%
(±)-Allyl 2,3-epoxypropyl ether
Formula: C₆H₁₀O₂
MW: 114.14 g/mol
Boiling Pt: 154 °C (760 mmHg)
Melting Pt: –100 °C
Density: 0.962 g/cm³
Flash Pt: 48 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005143
CAS Number: 106-92-3
EINECS: 203-442-4
UN: 2219
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4332 to 1.4352 (20 °C, 589 nm)
Water ≤0.2 % (K.F.)
GC ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR