α-Acetyl-γ-butyrolactone 99+%

Warning

Synonyms: 2-Acetyl-γ-butyrolactone3-Acetyldihydrofuran-2(3H)-one2-Acetylbutyrolactone

AC10249-2500 AC10249-1000
200016-256EA 30.5 USD
200016-256 200016-260
α-Acetyl-γ-butyrolactone 99+%
α-Acetyl-γ-butyrolactone
Formula: C₆H₈O₃
MW: 128.13 g/mol
Boiling Pt: 107…108 °C (5 mmHg)
Density: 1.18 g/cm³
Flash Pt: 140 °C
Storage Temperature: Ambient
MDL Number: MFCD00005394
CAS Number: 517-23-7
EINECS: 208-235-2

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4580 to 1.4610 (20 °C, 589 nm)
GC ≥99.0 %
Color scale ≤30 APHA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR