Acetaldehyde oxime (mixture of syn and anti) 99%

Warning

Synonyms: Acetaldoxime

AC10223-0250 AC10223-1000
200036-438EA 47.5 USD
200036-438 200017-956
Acetaldehyde oxime (mixture of syn and anti) 99%
Acetaldehyde oxime (mixture of syn and anti)
Formula: C₂H₅NO
MW: 59.07
Melting Pt: 44…46 °C
Density: 0.96 g/cm³
Flash Pt: 38 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002124
CAS Number: 107-29-9
EINECS: 203-479-6
UN: 2332
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to white Crystals or low melting solid or liquid
Infrared spectrum Conforms
Refractive index 1.4230 to 1.4270 (20 °C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR