1-Acetyl-4-(4-hydroxyphenyl)piperazine 98%

Warning

Synonyms: 1-(4-(4-Hydroxyphenyl)piperazin-1-yl)ethanone4-(4-Acetyl-1-piperazinyl)phenol

AC10034-0250 AC10034-0500
200033-854EA 42.3 USD
200033-854 200063-320
1-Acetyl-4-(4-hydroxyphenyl)piperazine 98%
1-Acetyl-4-(4-hydroxyphenyl)piperazine

Appearance: White to light pink Powder

Formula: C₁₂H₁₆N₂O₂
MW: 220.27 g/mol
Melting Pt: 180…185 °C
Storage Temperature: Ambient
MDL Number: MFCD00044905
CAS Number: 67914-60-7

Order Now

Specification Test Results

Appearance White to light pink Powder
Infrared spectrum Conforms
Melting point 180 to 185 °C
TLC ≤2 % Total sum impurities
UV transmittance ≥70 % (c=10, DMF, 400 nm, 1 cm cell)
Titration with HClO4 ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR