Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Membrane and Membrane Potential Dyes, Biotium

Supplier: Biotium
90047 90028 60037 80105 80104 60033 60032 60031 60030 70009 90010 70008 70007 61019 80012 30037 60028 61018 60027 61017 60026 61016 60025 61015 60024 61014 60023 61013 60022 61012 61011 60021 61010
Dyes for membrane potential measurements, vesicle fusion assays, and for binding to cell membranes or liposomes.

  • Study membrane fusion or permeability
  • Dyes for functional imaging of neurons
  • Membrane potential detection kits

The fluorescence of NBD is highly dependent on the environment; it is virtually nonfluorescent in water but becomes strongly green fluorescent in hydrophobic media

NBD-ethylenediamine can be readily coupled to activated carboxy groups or sulfonyl chlorides. ANTS can be used together with the fluorescent quencher DPX for the studies of membrane fusion or permeability. The mixture of ANTS-DPX is minimally fluorescent initially, but becomes increasingly more fluorescent upon dilution (i.e., membrane fusion or leakage).

APTS is a very useful green fluorescent dye for labeling glycoproteins or sugar molecules in general.

DPH is almost non-fluorescent in water, but it exhibits a strong increase in fluorescence after intercalation into membranes and can be used as a probe for viscosity, polarity and lipid order.  

NBD C6-ceramide has been used for following sphingolipid metabolism in cells, and as a selective stain for the Golgi apparatus in live and fixed cells. NBD C6-Sphingomyelin has been used for following sphingolipid metabolism in cells.
Biotin-AM3-25 is a conjugate of AM3-25 and biotin. Membranes labeled with the probe can subsequently couple to avidin or streptavidin.
DPA is a nonfluorescent anionic membrane dye. With its absorbance in the blue region of the visible spectrum, DPA can be used as a FRET quencher for a variety of fluorescent donor dyes.
ANEPPS dyes are fast responding membrane potential dyes. Di-4-ANEPPS has the problem of being internalized in cells over long-term, so it is mainly used for short-term experiments. Di-8-ANEPPS is more lipophilic, and the dye is better retained in the outer leaflet of cell plasma membranes and thus more suitable for long-term membrane potential studies. Di-8-ANEPPS is also more photostable and less phototoxic than Di-4-ANEPPS.DiBAC4(3) is a so-called translational membrane potential dye that redistributes within the cell membrane when membrane potential changes.
Di-2-ANEPEQ , Di-8-ANEPPQ and Di-12-ANEPPQ are fast-responding membrane potential dye. Di-2-ANEPEQ is highly water-soluble and is usually introduced into cells by microinjection. Di-8-ANEPPQ and Di-12-ANEPPQ are more hydrophobic than Di-2-ANEPEQ and useful for potential-sensitive retrograde labeling of neurons.
The styryl dye RH414, RH421, RH237 and RH795 are fast-responding potentiometric probes that are primarily used for functional imaging of neurons.
DiOC5(3) and DiOC6(3) are two of the most widely used carbocyanine dyes used for membrane potential measurements. DiOC2(3) has been used for measuring membrane potentials in bacteria. DiOC6(3) has been used to stain the ER in both live and fixed cells.
In the DPA/Tb3+ vesicle fusion assay, separate vesicle populations are loaded with TbCl3 or DPA. Fusion of the two types of vesicles results in fluorescence increase at 490 nm or 545 nm, with excitation at 276 nm. The Tb3+-DPA complex
10,000 times more fluorescent than free Tb3+. SDIP/Eu3+ as an alternative to DPA/Tb3+. The advantage is that SDIP/Eu3+ generates fluorescent emission with brighter intensity and much longer wavelength. DiO/DPA membrane potential detection kit is comprised of the green fluorescent membrane dye DiOC16(3) and the anionic quencher dye DPA.
The DiO/DPA system detects cytoplasmic membrane potential changes using FRET. DiO is a “stationary” donor while DPA acts as a mobile acceptor.




 

ORDER

Expand All / Collapse All
Description Size Ex/Em Supplier No. VWR Catalog Number Unit Price Quantity
DPX 500 mg 80012 89139-476 Each (500mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

NBD-Ethylenediamine 10 mg 466/535 nm 90047 89139-598 Each (10mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

ANTS 500 mg 353/520 nm 90010 89139-534 Each (500mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

APTS 10 mg 424/505 nm 90028 89139-564 Each (10mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

DPH 100 mg 350/420 nm 60021 10066-560 Each (100mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

BIOTIN-DHPE 10 mg 60022 89139-320 Each (10mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Biotin-X-DHPE 5 mg 60023 89139-322 Each (5mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Fluorescein-DHPE 5 mg 496/519 nm 60024 89139-324 Each (5mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

NBD-PE 10 mg 463/536 nm 60025 89139-326 Each (10mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Rhodamine-DHPE 5 mg 560/581 nm 60026 89139-328 Each (5mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Texas red-DHPE 1 mg 582/601 nm 60027 89139-330 Each (1mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

TRITC DHPE 1 mg 540/566 nm 60028 89139-332 Each (1mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

NBD C6-Ceramide 1 mg 466/536 nm 60030 89139-334 Each (1mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

NBD C6-Sphingomyelin 1 mg 466/536 nm 60031 89139-336 Each (1mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Biotin-AM3-25 1 mg 510/625 nm (480/600 nm in liposomes) 60032 89139-338 Each (1mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Octadecyl Rhodamine B 10 mg 556/578 nm 60033 89139-340 Each (10mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

DPA 25 mg 60037 89139-348 Each (25mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

DI-4-ANEPPS 5 mg 496/705 nm 61010 89139-352 Each (5mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

DIBAC4(3) 25 mg 490/516 nm 61011 89139-354 Each (25mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

DI-8-ANEPPS 5 mg 498/713 nm 61012 89139-356 Each (5mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

DI-2-ANEPEQ (also called JPW1114) 5 mg ~465/635 nm 61013 89139-358 Each (5mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

DI-8-ANEPPQ 5 mg 498/713 nm 61014 89139-360 Each (5mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

DI-12-ANEPPQ 5 mg 61015 89139-362 Each (5mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

RH414 5 mg 532/716 nm 61016 89139-364 Each (5mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

RH421 25 mg 515/704 nm 61017 89139-366 Each (25mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

RH237 5 mg 528/782 nm 61018 89139-368 Each (5mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

RH795 5 mg 530/712 nm 61019 89139-370 Each (5mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

DIOC5(3) 100 mg 482/497 nm 70007 89139-372 Each (100mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

DIOC2(3) 100 mg 482/497 nm 70008 89139-374 Each (100mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

DIOC6(3) 100 mg 484/501 nm 70009 89139-376 Each (100mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

DPA/Terbium for Membrane Fusion Assay 1 set 276/490 and 545 nm 80104 89139-510 Each Retrieving Restricted
SDIP/Europium for Membrane Fusion Assay 1 set 80105 89139-512 Each Retrieving Restricted
DIO/DPA Membrane Potential Detection Kit 1 kit 30037 89138-972 Each Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty