AlphaTec™ 66-677 Chemical Protection Flame Resistant Coveralls, Ansell
Supplier: Ansell
AlphaTec®
966445 966442 966441 966444
80093-563CS 15721.7 USD
80093-563 80093-561 80093-567 80093-569
AlphaTec™ 66-677 Chemical Protection Flame Resistant Coveralls, Ansell
Workwear
This lightweight, flexible, snag- and tear-resistant, stain-repellent and washable bib overall series provides outstanding chemical splash protection to a wide range of chemicals as well as excellent fire protection.
  • Offers HRC Level 2 arc flash protection
  • Increases chance of survival from exposure to thermal hazards
  • Does not sustain a flame and no melting or dripping during flammability testing
  • Chemical-resistant stitching and taped seams ensure superior performance
  • Tough and durable fabric
  • Anti-static properties
Breathable and lightweight Gore® chemical splash fabric transports moisture away from the body to reduce the risk of heat stress and provides all-day protection and comfort
Use in the chemical processing, general industry, and petrochemical industries. Uses include bulk chemical handling and transport, chemical plants and transportation, chemical processing, oil refining and petrochemicals, and handling specialty chemicals.
Full-coverage chemical protection with the added safety of fire resistance (Arc Hazard Rating 2).
Certifications: Certified as Liquid Splash Protective Clothing for Hazardous material incidents for escape use only in accordance with NFPA 1992. Meets Hazard Category 2 as defined by NFPA 70E. Material is certified to Arc (ATPV) rating of 9.0 cal/cm2 based upon ASTM F1891-02/ASTM 1959-04.