VWR® Bis-Tris Gel Buffer, 4X
Supplier: Avantor
VWRV1B1670-500ML
10154-300EA 111.33 USD
10154-300
VWR® Bis-Tris Gel Buffer, 4X
Bis-Tris Gel Buffer, 4X is used to prepare polyacrylamide gels for optimal separation of small- to medium-sized proteins under denaturing conditions
  • Faster run time-separate your proteins in less than 35 minutes
  • Obtain high resolution of small- to medium-sized proteins
  • Neutral pH of the gel minimizes protein modifications
  • Store cold
The neutral pH environment of the gel prepared with Bis-Tris Gel Buffer minimizes protein modifications.  Bis-Tris Gel Buffer, 4X can be used to prepare both separating and stacking gels with your standard protocol for polyacrylamide gels preparation. MES-SDS or MOPS-SDS buffer is then used as the running buffer to separate your proteins on a Bis-Tris gel. When using MES-SDS buffer, separation of proteins on a 10% Bis-Tris gel can be accomplished in less than 35 minutes.