Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

AlphaTec™ 56-001 PVC Aprons, Ansell

Supplier: Ansell Healthcare
56-001-35X45 B-45P 56001950003 950006
Wherever oil, grease and chemicals are present, Ansell aprons help workers keep clean and protected from harmful contact.

  • Excellent splash protection
  • Superior flexibility
  • Adjustable ties ensure optimal fit

These medium-duty aprons feature 7- to 8-millimeter PVC and have a sewn edge finish

Use in the food processing, food service, and chemical processing industries. Uses include food handling and preparation, industrial maintenance, and chemical processing.

They provide splash protection along with superior flexibility. Three-piece 28" ties and sealed grommets provide a durable, consistent and adjustable fit. Bib styles in two sizes with adjustable ties ensure optimal fit for a wide range of users.



 

ORDER

Expand All / Collapse All
Size Color Supplier No. VWR Catalog Number Unit Price Quantity
84W×112L cm (33×44") Blue 56001950003 52222-810 Case of 72 Retrieving
Pack of 12 Retrieving
89W×114L cm (33×45") Blue 56-001-35X45 76242-590 Case of 72 Retrieving
84W×137L cm (33×54") Blue 950006 80093-200 Case of 72 Retrieving
Pack of 12 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Material PVC
Thickness 8 mil

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty