SP Wilmad-LabGlass Economy NMR Sample Tubes, 5mm, SP Industries

Supplier: SP Industries
WG-5MMECNMY7
13501-528PK 16.32 USD
13501-528
SP Wilmad-LabGlass Economy NMR Sample Tubes, 5mm, SP Industries
Tubes NMR Tubes
Designed for routine use in all NMR spectrometers, Wilmad Economy NMR Tubes are guaranteed to fit tightly in any spinner turbine and have zero NMR background.

  • Best Ext.Ø tolerance in the industry and a 30% thicker wall than any competitors' product
  • Manufactured in an ISO 9001:2008 USA facility
  • Camber and concentricity values listed are the Total Indicator Reading (TIR)
  • 100% visual and physical inspection ensures quality, including physical dimensions and surface defects
  • Made from borosilicate glass; meets ASTM E438 Type 1 Class B standard, recognized as N51A

Recommended only for experiments with small organic molecules (Molecular Weight ~500) at ambient temperatures only.
Cooling/heating of these tubes may lead to breakage; for NMR experiments that involve cooling, heating, biological sample, multi-dimension, multi-nuclei or DNP techniques, refer to Wilmad Precision NMR Tubes.

Delivery information: Includes disposable cap.

Caution: Note: Caps are not recommended for use with samples dissolved in chloroform-d or acetone-d6.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR