Disposable Base Molds, Richard-Allan Scientific
Supplier: Richard-Allan Scientific
58327
CA15154-074
Disposable Base Molds, Richard-Allan Scientific
Moulds Moulds for Histology
Base molds for standard-sized cassettes and embedding rings.
  • Rigid plastic
  • Greater base support
  • Transparent
Disposable Base Molds are designed for use with all standard-sized cassettes and embedding rings. They are molded from rigid plastic, and the block design provides greater base support and a smaller cutting surface. Cleaning associated with metal molds is not required and the transparency facilitates tissue orientation.