E.Z.N.A.® Plant RNA Midiprep Kit, Omega Bio-tek
R6628-00 R6628-01 R6628-00
101414-828EA 37.83 USD
CA101414-828 101414-828 CA101319-210
E.Z.N.A.® Plant RNA Midiprep Kit, Omega Bio-tek
The E.Z.N.A.® Plant RNA Midiprep Kit provides a convenient and rapid method for the isolation of total RNA from a variety of plant samples.
  • RNA isolation under 60 minutes
  • Optimised buffer system ensure the reproducible results
  • No organic extractions
  • Purified RNA suitable for most downstream applications
The kit includes shredding/homogenizing units to efficiently remove cell debris and simultaneously homogenize the lysate
Purified RNA has Abs260/Abs280 ratios of 1.8-2.0 and is suitable for applications like RT-PCR, northern analysis, differential display and Poly A+ RNA selection.
In combination with HiBind® RNA spin columns, this permits purification of high quality RNA from as much as 500 mg of tissue.