Prionex®, BioVision
Supplier: BioVision
AB286838-100ML
10835-312EA 528.77 USD
10835-312
Prionex®, BioVision
Protein Purification Protein Purification Reagents
An effective BSA alternate, serving as a protein stabilizer, blocking agent and media additive.
  • Non-toxic and non-antigenic
  • Filterable through a 0.2 µm membrane
  • Heat-stable
  • Free from nucleic acids, polysaccharides, additives and lipids
  • No risk of BSE, MCD and HIV infective agents
An effective BSA alternate, serving as a protein stabilizer, blocking agent and media additive. Due to the chemical nature and the standardized quality, Prionex® can be used for multiple purposes such as an inert protein stabilizer in many applications, a protective additive in cell culture and a blocking agent.