VascuLife® VEGF Endothelial Medium Kit, Lifeline Cell Technology
Supplier: Lifeline Cell Technology
LS-1020 LM-0002 LL-0003
102970-762EA 153.08 USD
102970-762 102970-942 102970-922
VascuLife® VEGF Endothelial Medium Kit, Lifeline Cell Technology
Cell Culture Media
VascuLife® VEGF is optimized for the culture of human endothelial cells, and supports the rapid proliferation of these cells in a 5% serum environment.
  • Optimized for the culture of human endothelial cells
  • Supports rapid cell proliferation
  • Maintain excellent cell morphology
The VascuLife® VEGF Medium Complete Kit includes: VascuLife® Basal Medium (475 mL) and VascuLife® VEGF LifeFactors® Kit. VascuLife® contains no antimicrobials and no phenol red. VascuLife® VEGF contains Vascular Endothelial Growth Factor. VascuLife® VEGF Medium grows human endothelial cells at rates that meet or exceed other commercially available serum-containing media while maintaining excellent cell morphology.
Ordering information: The VascuLife® VEGF LifeFactors® Kit includes: rh VEGF 0.5 mL, rh EGF 0.5 mL, rh FGF basic 0.5 mL, rh IGF-1 0.5 mL, Ascorbic Acid 0.5 mL, Hydrocortisone Hemisuccinate 0.5 mL, Heparin Sulfate 0.5 mL, L-Glutamine 25 mL, FBS 10 mL, Gentamicin and Amphotericin B 0.5 mL.
VascuLife® Medium Complete Kits contain VascuLife® Basal Medium and the associated LifeFactors® Kit. VascuLife® Basal Medium requires the appropriate LifeFactors®.