Shandon™ Finesse™ Cold Point Light Source, Thermo Scientific
Supplier: Thermo Scientific
77510252 77510261 77510252 77510261
89238-804EA 5248.24 USD
89238-804 CA89238-802 CA89238-804 89238-802
Shandon™ Finesse™ Cold Point Light Source, Thermo Scientific
Lights Cold Light Sources
Direct two powerful beams of light to the specimen with the Shandon™ Finesse Cold Point Light Source.
  • Fiber optic lights do not affect specimen temperature
  • Gooseneck shafts provide maximum flexibility