Would you like to log in again?
Droplet® Personal Lancet, HTL-Strefa
Supplier: HTL-STREFA INC.
7206 7167 7106
10220-652BX 6.33 USD InStock
10220-652 10220-647 10220-645
Droplet® Personal Lancet, HTL-Strefa
Blood Lancets
Droplet® Personal Lancets are single-use devices that allow users to personalize lancing penetration depths
Perfectly ground ultra-sharp gamma sterilized needles are electro-polished and silicone-coated for maximum comfort and quick healing. The 33 gauge lancets feature ComfortMax® Technology, which offers the thinnest needle and highest comfort. The 30 gauge lancets are intended for the delicate skin of children or elderly. The 28 gauge lancets are suggested for use with tough or calloused skin.
Designed with a gentle silicone-coated needle, they provide smooth, vibration-free puncture for maximum comfort.
Certifications: Certified gamma sterilization.
Ordering information: Orders should be placed for case quantities.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty