EasyMIX® Blender, bioMerieux
Supplier: bioMérieux
AESAP1068
97059-210EA 6119.66 USD
97059-210
EasyMIX® Blender, bioMerieux
Homogenizers Laboratory Paddle Blenders
The EasyMIX® Blender provides high quality sample preparation by combining superior blending with a fast speed.
  • High quality sample preparation
  • Accurate electronic timer
  • Stores homogenization times
  • Digital control display
  • Programmable timer
  • Wash-down keypad
  • Easy-to-clean with fully opening door
  • Electronic protection of the motor and paddle mechanisms
  • Stainless steel chamber and blending paddles
After the dilution, blending is an essential step for the quality of microbiological analysis. An accurate electronic timer sets and stores the time for homogenization. Time savings of up to 50% can be made in this area with no compromise of homogenization of sample or distribution of bacteria.
EasyMIX is easy to maintain and clean as all the components, including the two paddles, are made of stainless steel and easily accessible. Optional waste drawer and/or plexiglass door.