Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Smasher™ High-Performance Blender/Homogenizer, bioMerieux

Supplier: bioMérieux
AESAP1064
The Smasher™ High-Performance Blender/Homogenizer provides blending at a very low noise level for perfectly homogenized enrichment broth.

  • Digital control, keypad and LCD display
  • Elapsed time bargraph display
  • Adjustable speed
  • Blending capacity of 80 to 400 mL bags
  • Removable stainless steel paddles
  • Integrated waste drawer
  • Leak-proof blending chamber
  • Protection of the motor and machinery
  • Electronic circuit-breaker
  • Door activated start/stop

Discover the unique features of this advanced paddle blender for microbiology laboratories. Smasher™ provides high-performance blending at a very low noise level. Thanks to the patented 'SMASH effect', it crushes the sample in record time and then automatically adjusts its mixing speed for a perfectly homogenized enrichment broth. Operation is very quiet, producing 19 to 30 times less noise than other peristaltic blenders. Total blending time is reduced to 15 sec.

Smasher avoids jams thanks to the automatic control of difficult samples; there is no need to reposition the bag. It senses any sample that prevents full travel of its paddles and automatically returns to its starting position from where it resumes the task, without operator intervention.

Smasher is the first peristaltic blender featuring a leak-free and corrosion-free blending chamber, with an integrated waste drawer. For easy cleaning, the leak-proof and corrosion-resistant blending chamber has rounded corners and is easily accessible via the wide-opening door. All chamber and housing surfaces are extremely robust and resistant to chemical damage from cleaning agents. Paddles can be easily removed within two seconds without any tools. The unique integrated waste drawer ensures that spills are contained. Can be tilted back at 45° for improved access.




 

ORDER

Expand All / Collapse All
Description Capacity Dimensions Speed Range Weight Supplier No. VWR Catalog Number Unit Price Quantity
Smasher™ Blender/Homogenizer 400 mL 40W x 33H x 38D cm (16 x 13 x 15”) 500 - 620 strokes/min 21 kg (46 lbs.) AESAP1064 97059-208 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty