Potassium hydrogen phthalate 99.95-100.05% ACS
Supplier: VWR
Synonyms:
KHP
, Potassium phthalate monobasic
, Potassium biphthalate
, Phthalic Acid Monopotassium Salt
, Phthalic acid potassium salt
VWRVN983-100G
10791-800CS
546.6
USD
10791-800
Potassium hydrogen phthalate 99.95-100.05% ACS
Potassium hydrogen phthalate
Formula:
HOOCC₆H₄COOK MW: 204.22 g/mol Melting Pt: 295…300 °C Density: 1.6362 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00013070 CAS Number: 877-24-7 EINECS: 212-889-4 Merck Index: 12,07773 |
Specification Test Results
Assay (dried basis) | 99.95% - 100.05% |
Chlorine Compounds |
|
Heavy Metals (as Pb) |
|
Insoluble Matter |
|
Iron |
|
pH (0.05M, Water) @25°C | 4 - 4.02 |
Sodium |
|
Sulfur Compounds |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...