Bovine Serum Albumin (BSA), CML-modified
Supplier: Cell Biolabs
STA-314
102964-890EA 235 USD
102964-890
Bovine Serum Albumin (BSA), CML-modified
Proteins and Peptides
Nε-(carboxymethyl) lysine (CML) modified BSA is provided at a concentration of 1 mg/mL in PBS.