PYREX® Jacketed Kuderna-Danish Concentrator Tube with Serrated Tubulations, Corning®
2158-10JTO 2158-1JTO
89091-110CS 242.52 USD
89091-110 89091-222
PYREX® Jacketed Kuderna-Danish Concentrator Tube with Serrated Tubulations, Corning®
These innovative PYREX® jacketed Kuderna-Danish concentrator tubes circulate hot water through the jacket to boil the solvent during extraction/concentration
The bottom portion of the tube is never heated, so samples won't boil dry. The concentration automatically stops when the solvent sinks below the jacket. Lab technician time is saved by eliminating the need to watch evaporations closely to avoid boiling dry. Serrated tubulations allow for easy tubing connections.