Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

IntelliCAL™ PHC729 Red Rod pH Electrode, 1 meter cable, Hach

Supplier: Hach
PHC72901
IntelliCAL PHC729 Red rod pH electrode for surface measurements

  • High performance Red Rod technology now available for the digital HQD platform
  • Red Rod technology provides fast, accurate and reproducible measurements even in samples with fluctuating temperatures
  • Red Rod reliability when measuring challenging samples over wider temperature ranges
  • IntelliCAL™ digital probes provide ultimate traceability in measurement history
  • IntelliCAL™ digital probes can be moved between meters without the need to re-calibrate or re-enter measurement settings

Symmetry between internal and external Red Rod reference elements ensures that the iso pH is the same as the zero pH. Red Rod technology also provides a stable signal by protecting the reference elements from light interference, and the saturated KCl salt-bridge aids in creating a stable liquid junction potential.



 

ORDER

Expand All / Collapse All
Description Supplier No. VWR Catalog Number Unit Price Quantity
IntelliCAL™ PHC729 Red Rod pH Electrode with 1 m Cable, for Surface Measurements PHC72901 10146-006 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Accuracy ±0.01 pH
Probe type Standard
Range 0 to 12 pH
Cable Length 1 m
Ø 12 mm (1/2")
Temperature Range 0 to 100 °C
Optic Range
Body Material Glass
Water Resistance IP65
Reference Type Red Rod
Filling Solution Sat. KCl
Junction Annular Ring

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty