Histamine Assay Kit, Cell Biolabs
Supplier: Cell Biolabs
AKR-360
102963-674EA 415 USD
102963-674
Histamine Assay Kit, Cell Biolabs
Cell Biolabs' Histamine Assay Kit detects total histamine from a variety of food samples such as meat, seafood, fish meal, milk and wine
This kit uses a colorimetric probe to measure histamine levels. Reduction of the probe yields color development proportional to the histamine levels in the sample. After a quick one hour incubation, absorbance at 450nm is read in a standard microplate reader, and histamine levels are calculated based on a histamine standard curve. The Histamine Assay Kit can detect as little as 0.8 µg/ml (0.8 ppm) histamine.
Histamine is naturally occurring in food, with high concentrations associated with spoiled and fermented foods. Exposure to high levels of histamine through the ingestion of food can cause symptoms similar to an allergic response.