Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

HxSil C18 HPLC Guard Columns, Cation Exchange, Hamilton

Supplier: HAMILTON COMPANY
79288 79290 79294 79914 79917 79916 79919
Hamilton guard columns protect analytical, semi-prep and preparative HPLC columns

  • Protect columns
  • Remove particulates
  • Modular design
  • Reduces dead volume
  • Prevents column band broadening

Cartridge holders for analytical columns are available in stainless steel and PEEK materials. Semi-prep/preparative holders are available only in stainless steel. The design of the cartridge holders is optimized to reduce dead volume and prevent extra column band broadening. Cartridge holders are reusable indefinitely.

They also remove particulate contaminants and highly adsorptive compounds from samples, prolonging column life. The designs are modular and cartridge replacement is easy and tool-free.




 

ORDER

Expand All / Collapse All
Column Format Column Hardware Phase Packing Material Particle Size Supplier No. VWR Catalog Number Unit Price Quantity
Analytical Stainless Steel PSDVB/Sulfonic Acid PRP-X200 PSDVB/Sulfonic Acid 10 µm 79288 10836-954 Each Retrieving
Analytical Stainless Steel PSDVB/Sulfonic Acid PRP-X400 PSDVB/Sulfonic Acid 7 µm 79290 10836-958 Each Retrieving
Analytical Stainless Steel PRP-X800 PSDVB/Itaconate 7 µm 79294 10836-960 Each Retrieving
Analytical Stainless Steel Poly(methacrylamidopropryl Trimethylammonimu chloride) PRP-X500 PSDVB/Sulfonic Acid 12 - 20 µm 79919 10836-964 Each Retrieving
Analytical Stainless Steel PSDVB/Sulfonic Acid PRP-X400 PSDVB/Sulfonic Acid 12 - 20 µm 79917 10836-970 Each Retrieving
Analytical Stainless Steel PSDVB/Sulfonic Acid PRP-X200 PSDVB/Sulfonic Acid 12 - 20 µm 79914 10836-982 Each Retrieving
Analytical Stainless Steel PSDVB/Sulfonic Acid PRP-X400 PSDVB/Sulfonic Acid 12 - 20 µm 79916 10836-986 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Description 10836-958 10836-982 10836-986 10836-954 10836-964 10836-960 10836-970
Application Glyphosate Organic/Inorganic Cations Glyphosate Organic/Inorganic Cations Carbohydrates Mono-Divalent Organic/Inorganic Cations - Transition Metals Carbohydrates

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty