Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

IL-17F, rat recombinant, BioVision

Supplier: BioVision
4178F-1000 4178F-25 4178F-100
>98% Pure active Recombinant Rat Interleukin-17F (IL-17F)

IL-17F is fully bioactive when compared to a known standard. The ED₅₀ as determined by the dose-dependant induction of IL-6 in NHDF is 1.94 ng/ml.

Recombinant Rat IL-17F produced in E.coli is a homodimeric, non-glycosylated polypeptide chain containing a total of 133 amino acids and having a molecular mass of 14.9 kDa.




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
25 μg 4178F-25 10006-332 Each (25µG) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

100 μg 4178F-100 10006-328 Each (100µG) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

1 mg 4178F-1000 10006-330 Each (1mg) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Protein/Peptide Name IL17F
Protein Synonyms rat IL-17F, rat IL-17, IL17F, IL 17F, IL-17, rr-IL-17, r-IL-17F IL-17F, r-IL-17F, rr-IL-17, IL-17, IL 17F, IL17F IL17F, r-IL-17F, rat IL-17, rat IL-17F, IL-17, IL 17F, rr-IL-17
Protein/Peptide Type Recombinant
Species Rat
Source E. coli
Purity ≥98%
Endotoxin Content <0.01 ng/μg
Molecular Weight 14.9 kDa
Formulation Lyophilized with no additives
Reconstitution Instructions Reconstitute in sterile H₂O to a concentration of 0.1 mg/ml.
Protein Type Recombinant
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty