Rat Recombinant IL17E (from E. coli)
Supplier: BioVision
4178E-1000 4178E-10 4178E-100
10006-326EA 5884.18 USD
10006-326 10006-324 10006-322
Rat Recombinant IL17E (from E. coli)
>95% Pure active Recombinant Rat Interleukin-17E (IL-17E)
Rat IL-17E (IL-17E), is a proinflammatory cytokine member of a six-species family of proteins (IL-17A-17F). Rat IL-17E protein is a homodimer consisting of two 145 amino acids peptides. Recombinant rat IL-17E produced in E.coli is a non-glycosylated disulfide-joined homodimer having a molecular mass of 35.5 kDa.