Anti-DRD4 Mouse Monoclonal Antibody [clone: 625CT1.5.4]

Supplier: Abcepta
AM2167B
89514-468EA 636.94 USD
89514-468
Anti-DRD4 Mouse Monoclonal Antibody [clone: 625CT1.5.4]
Antibodies
Western Blot: 1:100~250,

Type: Primary
Antigen: DRD4
Clonality: Monoclonal
Clone: 625CT1.5.4
Conjugation: Unconjugated
Epitope: Center
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR