Anti-RPB3 Mouse Monoclonal Antibody [clone: 1Y26]

665003
10019-924EA 309.26 USD
10019-924
Anti-RPB3 Mouse Monoclonal Antibody [clone: 1Y26]
Type: Primary
Antigen: RPB3
Clonality: Monoclonal
Clone: 1Y26
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Yeast (Saccharomyces cerevisiae)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR