Anti-GPATCH11 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP18182A
89516-876EA 636.79 USD
89516-876
Anti-GPATCH11 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:1000

Type: Primary
Antigen: GPATCH11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human, Hamster, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR