|
![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | SH2D5 |
Antigen Name | SH2 Domain Containing 5 |
Reactivity | Mouse |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Antibody Type | Primary |
Host | Rabbit |
Western Blot | Yes |
ELISA | Yes |
Amino Acid Number | 200 to 230 |
Epitope | Center |
Concentration | 0.5mg/ml |
Immunogen | This Mouse Sh2d5 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 200-230 amino acids from the Central region of mouse Sh2d5. |
Molecular Weight | 38.174 kDa |
Purification | Protein A & Peptide affinity purification |
Sequence | PESEGRGGTRHIRC |
Storage Buffer | PBS with 0.09% (W/V) sodium azide, pH7.4 |
Storage Temperature | Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C in small aliquots to prevent freeze-thaw cycles |
Shipping Temperature | 4°C |