|
![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | C5AR2 |
Antigen Name | Complement Component 5a receptor 2 |
Reactivity | Human |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Antibody Type | Primary |
Host | Rabbit |
Western Blot | Yes |
ELISA | Yes |
Antigen Synonyms | GPR77, GPF77, C5L2 |
Amino Acid Number | 297 to 325 |
Gene ID | 27202 |
Epitope | C-Terminal |
Concentration | 0.29mg/ml |
Immunogen | This GPR77 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 297-325 amino acids from the C-terminal region of human GPR77. |
Molecular Weight | 36.08 kDa |
Purification | Protein A & Peptide affinity purification |
Sequence | RESQGQDESVDSKKC |
Storage Buffer | PBS with 0.09% (W/V) sodium azide, pH7.4 |
Storage Temperature | Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C in small aliquots to prevent freeze-thaw cycles |
Shipping Temperature | 4°C |