Anti-CAMSAP3 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP18323A
89516-448EA 636.94 USD
89516-448
Anti-CAMSAP3 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:1000

Type: Primary
Antigen: CAMSAP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR