Anti-SH3BP4 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP19380C
89514-862EA 636.79 USD
89514-862
Anti-SH3BP4 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:1000

Type: Primary
Antigen: SH3BP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR