Anti-SEMA4A Mouse Monoclonal Antibody [clone: 5E3/SEMA4A]

Supplier: BioLegend
148402 148402
10756-786EA 297.94 USD
CA10756-786 10756-786
Anti-SEMA4A Mouse Monoclonal Antibody [clone: 5E3/SEMA4A]
Antibodies
Purified anti-mouse/human SEMA4A [5E3/SEMA4A]; Isotype: Mouse IgG1, λ; Reactivity: Mouse, Human; Apps: FC, ELISA; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: SEMA4A
Clonality: Monoclonal
Clone: 5E3/SEMA4A
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, λ
Reactivity: Mouse;Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR