Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PTEN Mouse Monoclonal Antibody () [clone: 4C11A11]

Supplier: Biolegend
Purified anti-PTEN [4C11A11]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Mouse; Apps: WB, IF; Size: 100 μg

Each lot of this antibody is quality control tested by Western blotting. Western blotting, suggested working dilution(s): Use 0.5-2.5 μg/ml antibody dilution buffer for each mini-gel. For <a title="View Protocol" href="" target="blank">immunofluorescence microscopy</a>, a concentration range of 2.5-5 μg/ml is recommended. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: PTEN
Clonality: Monoclonal
Clone: 4C11A11
Host: Mouse
Isotype: Mouse IgG2b, κ
Reactivity: Human;Mouse

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 μg 655002 10763-522 Each (100µG) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen Symbol PTEN
Antigen Name PTEN
Reactivity Mouse, Human
Clonality Monoclonal
Antibody Type Primary
Clone 4C11A11
Host Mouse
Isotype Mouse IgG2b, κ
Western Blot Yes
ImmunoFluorescence Yes
Antigen Synonyms MMAC1, BZS, TEP1, MHAM1, GLM2, PTEN1
Gene ID 5728
Concentration 0.50 mg/mL
Immunogen Partial human PTEN recombinant protein (303-403aa)
Purification Affinity purified.
Form This antibody is provided in phosphate-buffered solution, pH 7.2, containing 0.05% sodium azide.
Storage Temperature Upon receipt, store undiluted between 2°C and 8°C.
Shipping Temperature Ambient RT
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty