Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-XCR1 Mouse Monoclonal Antibody () [clone: ZET]

Supplier: Biolegend
148202
Purified anti-mouse/rat XCR1 [ZET]; Isotype: Mouse IgG2b, κ; Reactivity: Mouse, Rat; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: XCR1
Clonality: Monoclonal
Clone: ZET
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, κ
Reactivity: Mouse;Rat




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 μg 148202 10756-636 Each (100µG) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol XCR1
Antigen Name XCR1
Reactivity Rat, Mouse
Clonality Monoclonal
Antibody Type Primary
Clone ZET
Host Mouse
Isotype Mouse IgG2b, κ
Antigen Synonyms GPR5, CCXCR1, mXCR1, Lymphotactin receptor, SCM1 receptor, XC chemokine receptor 1
Gene ID 23832, 301086
Concentration 0.50 mg/mL
Purification The antibody was purified by affinity chromatography.
Form Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
Storage Temperature The antibody solution should be stored undiluted between 2°C and 8°C.
Shipping Temperature Ambient RT
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping