Anti-VEGFR-3 Rat Monoclonal Antibody (LEAF) [clone: AFL4]

140904 140904
140904-BLEA 311.22 USD
140904-BL CA140904-BL
Anti-VEGFR-3 Rat Monoclonal Antibody (LEAF) [clone: AFL4]
LEAF™ Purified anti-mouse VEGFR-3 (FLT-4) [AFL4]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: IHC, Block, ELISA, IP, WB; Size: 500 μg

Each lot of this antibody is quality control tested by <a href="http://www.biolegend.com/media_assets/support_protocol/BioLegend_IHC_Frozen_121511.pdf">immunohistochemical staining</a> of mouse frozen tissue sections.

Type: Primary
Antigen: VEGFR-3
Clonality: Monoclonal
Clone: AFL4
Conjugation: LEAF
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR