Anti-VEGFR-3 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 9D9F9]

Supplier: BioLegend
356203 356203 356204 356204
10759-954EA 165.11 USD
10759-954 10759-956 CA10759-956 CA10759-954
Anti-VEGFR-3 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 9D9F9]
Antibodies
PE anti-human VEGFR-3 (FLT-4) [9D9F9]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: VEGFR-3
Clonality: Monoclonal
Clone: 9D9F9
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR