Anti-VEGF-A Mouse Monoclonal Antibody [clone: L308D10]

Supplier: BioLegend
662702 662702
10763-626EA 124.09 USD
10763-626 CA10763-626
Anti-VEGF-A Mouse Monoclonal Antibody [clone: L308D10]
Antibodies
Purified anti-VEGF-165 [L308D10]; Isotype: Mouse IgG1; Reactivity: Human; Apps: IP; Size: 100 μg

<span>Each lot of this antibody is quality control tested by </span><a href="http://www.biolegend.com/media_assets/support_protocol/BioLegend_Immunoprecipitation_protocol_121511.pdf">immunoprecipitation</a><span>. This antibody can be used at 2 - 10 μg/mL</span><span> for immunoprecipitation. It is recommended that the reagent be titrated for optimal performance for each application.</span>

Type: Primary
Antigen: VEGF-A
Clonality: Monoclonal
Clone: L308D10
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR