Anti-TSLP Goat Polyclonal Antibody [clone: Poly5159]

Supplier: BioLegend
515901 515901
10761-706EA 376.9 USD
CA10761-706 10761-706
Anti-TSLP Goat Polyclonal Antibody [clone: Poly5159]
Antibodies
Purified anti-human TSLP [Poly5159]; Isotype: Goat Polyclonal Ig; Reactivity: Human; Apps: ELISA Capture, WB; Size: 100 μg

Each lot of this antibody is quality control tested by ELISA assay.
For use as an ELISA capture antibody, a concentration range of 0.5-2 μg/ml is recommended. It is recommended that the reagent be titrated for optimal performance.
For use in Western blotting, a suggested working concentration of 0.2-1 μg/mL is recommended.

Type: Primary
Antigen: TSLP
Clonality: Polyclonal
Clone: Poly5159
Conjugation:
Epitope:
Host: Goat
Isotype: Goat Polyclonal Ig
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR