Anti-ALPL Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: TRA-2-54/2J]

Supplier: BioLegend
358805 358806 358806 358805
10760-474EA 428 USD
CA10760-474 10760-474 10760-472 CA10760-472
Anti-ALPL Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: TRA-2-54/2J]
Type: Primary
Antigen: ALPL
Clonality: Monoclonal
Clone: TRA-2-54/2J
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR