Anti-TRA-2-49 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: TRA-2-49/6E]

Supplier: BioLegend
358706 358706 358705
10760-462EA 437.14 USD
10760-462 CA10760-460 CA10760-462
Anti-TRA-2-49 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: TRA-2-49/6E]
Antibodies
Alexa Fluor® 488 anti-human TRA-2-49 [TRA-2-49/6E]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: TRA-2-49
Clonality: Monoclonal
Clone: TRA-2-49/6E
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR