Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TNF Mouse Monoclonal Antibody (APC (Allophycocyanin)/Cy7®) [clone: MAb11]

Supplier: Biolegend
502943 502944
Type: Primary
Antigen: TNF
Clonality: Monoclonal
Clone: MAb11
Conjugation: APC (Allophycocyanin)/Cy7®
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Chimpanzee, Baboon, Cynomolgus, Rhesus, Pig, Pigtailed Macaque, Sooty Mangabey




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 Tests 502944 10761-354 Each Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

25 tests 502943 10761-352 Each Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol TNFA
Antigen Name Tumor Necrosis Factor Alpha
Reactivity Pig, Pigtailed Macaque, Chimpanzee, Baboon, Cynomolgus Monkey, Human, Rhesus Monkey, Sooty Mangabey Human, Pigtailed Macaque, Rhesus Monkey, Cynomolgus Monkey, Chimpanzee, Sooty Mangabey, Baboon, Pig
Conjugation APC (Allophycocyanin)/Cy7®
Clonality Monoclonal
Antibody Type Primary
Clone MAb11
Host Mouse
Isotype IgG1 kappa
Flow Cytometry Yes
Antigen Synonyms TNF superfamily, Tnfsf1a, TNFalpha, monocyte-derived, TNF-a, tumor necrosis factor-alpha, member 2, tumor necrosis factor alpha, TNFSF2, cachectin, macrophage-derived, tumor necrosis factor ligand superfamily member 2, TNF, TNF-alpha, Tnfa, DIF, APC1 protein APC1 protein, monocyte-derived, TNF-alpha, TNF, tumor necrosis factor-alpha, cachectin, DIF, TNFSF2, macrophage-derived, member 2, tumor necrosis factor alpha, TNFalpha, TNF superfamily, tumor necrosis factor ligand superfamily member 2, TNF-a, Tnfsf1a, Tnfa
Gene ID 7124
Immunogen E. coli -expressed, recombinant human TNF-alpha
Purification The antibody was purified by affinity chromatography and conjugated with APC/Cy7 under optimal conditions. The solution is free of unconjugated APC/Cy7 and unconjugated antibody.
Form Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
Storage Temperature The antibody solution should be stored undiluted between 2°C and 8°C, and protected from prolonged exposure to light. Do not freeze.
Shipping Temperature Ambient RT
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty