Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TNF Rat Monoclonal Antibody [clone: MP6-XT22]

Supplier: Biolegend
Type: Primary
Antigen: TNF
Clonality: Monoclonal
Clone: MP6-XT22
Conjugation:
Epitope:
Host: Rat
Isotype: IgG1 kappa
Reactivity: Mouse



Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µg 506335 10761-964 Each (100µG) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol TNFA
Antigen Name Tumor Necrosis Factor Alpha
Reactivity Mouse
Clonality Monoclonal
Antibody Type Primary
Clone MP6-XT22
Host Rat
Isotype IgG1 kappa
ELISA Yes
Gel Shift Yes
Antigen Synonyms cachectin, TNF-alpha, Tnfa, TNFalpha, TNF superfamily, DIF, TNFSF2, TNF, APC1 protein, macrophage-derived, member 2, tumor necrosis factor alpha, tumor necrosis factor-alpha, Tnfsf1a, TNF-a, tumor necrosis factor ligand superfamily member 2, monocyte-derived
Gene ID 21926
Concentration 1.0 mg/ml
Immunogen E. coli -expressed, recombinant mouse TNF-alpha
Purification The antibody was purified by affinity chromatography.
Form Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and EDTA.
Storage Temperature The antibody solution should be stored undiluted between 2°C and 8°C.
Shipping Temperature Ambient RT
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty