Anti-TAS2R7 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP16822B
89519-308EA 636.79 USD
89519-308
Anti-TAS2R7 Rabbit Polyclonal Antibody
Antibodies
  • Western Blot: 1:1000

Type: Primary
Antigen: TAS2R7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR